Orders LinkContactUsLinkComponent
PYREX® Reusable Volumetric Pipettes
   Sustainable Options Available
Supplier:   Corning
PYREX® Reusable Volumetric Pipettes
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Reusable Volumetric Pipettes
3 Options Available Below
Product Details & Documents
These reusable pipettes are designed primarily for schools and laboratories where the Class A tolerance is not required.
  • Larger tip openings for faster delivery
  • Colour-coded
  • Coloured graduations
  • Accuracy tolerances are twice those specified for Class A volumetric ware
They are constructed to the same wall thickness and length as standard Class A volumetric pipettes but have larger tip openings for faster delivery. Primarily due to the faster delivery, accuracy tolerances are twice those specified for Class A volumetric ware. Reference: ASTM E-969.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470211-614
  • 470211-610
  • 470211-612
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • Transfer pipette
    5 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $27.30
    $27.30
    Log in for availability
    Case of 12
    Case of 12 - $327.60
    $327.60
    Each
       $27.30
    Case of 12
       $327.60
    Supplier Number: 7102-5
    Specifications:
    More
    Specifications
    Cat. No.
    470211-614
    Environmentally Preferable
    Environmental certification
  • Transfer pipette
    10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $30.20
    $30.20
    Log in for availability
    Case of 12
    Case of 12 - $362.40
    $362.40
    Each
       $30.20
    Case of 12
       $362.40
    Supplier Number: 7102-10
    Specifications:
    More
    Specifications
    Cat. No.
    470211-610
    Environmentally Preferable
    Environmental certification
  • Transfer pipette
    25 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $45.40
    $45.40
    Log in for availability
    Case of 12
    Case of 12 - $544.80
    $544.80
    Each
       $45.40
    Case of 12
       $544.80
    Supplier Number: 7102-25
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-612
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact