Towering Toothpick Disaster Lab Activity
Catalog # 470016-768
Supplier: Avantor
Specifications
- Description:Towering Toothpick Disaster Lab Activity
- No. of Lab Groups:15
- Cat. no.:470016-768
- Supplier No.:WARD470016-768
Specifications
About this item
Using basic building materials, students design and build a three-story, earthquake resistance structure.
- Materials for 15 groups
- Teacher-Developed and Tested
- Students Apply Geology Knowledge to Solve a Real-World Problem
In an optional extension activity, they can also design and build a base isolator.
The teacher-designed activity comes with instructions and enough materials for 15 setups.
Accessories information: To further their learning, students can test whether their structure can withstand an earthquake with the Shake, Rattle, and Roll Earthquake Board Activity Set, available separately.