Tourmaline (Rubellite) Mineral Display
Catalog # 470154-890
Supplier: ADAM MOORE MO
Specifications
- Description:Tourmaline (Rubellite) Mineral Display, 2" to 2" x 3"
- Cat. no.:470154-890
- Supplier No.:3495625
Specifications
About this item
Brazil
These gemmy pink tourmaline crystal masses are showcased in a contrasting quartz matrix. Average size: 2" x 2" to 2" x 3". Limited Edition. Brazil.