Orders LinkContactUsLinkComponent
Thymus, Thin Section Slide
Thymus, Thin Section Slide
Thymus, Thin Section Slide, Thin Section, Hematoxylin and Eosin
Catalog # 470183-676
Supplier:   Avantor
Thymus, Thin Section Slide
Catalog # 470183-676
Supplier:  Avantor
Thymus, Thin Section Slide
Thymus, Thin Section Slide
Thymus, Thin Section Slide, Thin Section, Hematoxylin and Eosin
Price
19,35 $
Log in for availability
Each
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows Hassall’s corpuscles and other details of the organ.
Specifications
  • Description:
    Thymus, Thin Section Slide
  • Stain:
    Hematoxylin and Eosin
  • Orientation:
    Thin Section