ContactUsLinkComponent
Testis, Showing Spermatogenesis
Testis, Showing Spermatogenesis
Catalog # 470183-698
Supplier:  Avantor
undefined
Testis, Showing Spermatogenesis
Catalog # 470183-698
Supplier:  Avantor
Specifications
  • Description:
    Testis, Spermatogenesis
  • Cat. no.:
    470183-698
  • Stain:
    Iron Hematoxylin
  • Supplier No.:
    WARD470183-698
  • Orientation:
    Section
Specifications
About this item
Extremely Thin, Plastic Embedded Tissue For Greater Viewing Clarity.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rodent. Details of meiotic division clearly shown.