ContactUsLinkComponent
Disposable Polygon Spinbar Magnetic Stirring Bars
Disposable Polygon Spinbar Magnetic Stirring Bars
Catalog # 470153-810
Supplier:  Bel-Art
undefined
Disposable Polygon Spinbar Magnetic Stirring Bars
Catalog # 470153-810
Supplier:  Bel-Art
Supplier Number:  37112-6016
Specifications
  • Description:
    15/8" x 5/16"
  • Cat. no.:
    470153-810
  • Supplier No.:
    F371125016
Specifications
About this item
The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.