Kemtec® Tallquist Hemoglobin Scale
Tallquist Hemoglobin Scale
Catalog # 470023-872
Supplier: Kemtec
Supplier #: 12-136
Jump to:
Product Details & Documents
Standard hemoglobin reference scale.
- A fast test to determine anemia levels
- Paper for 150 tests
The Tallquist hemoglobin scale allows a quick assessment of a patient's hemoglobin levels. Allow blood to absorb into one of the test papers and compare the blood to the color scale to determine the concentration of hemoglobin in blood under normal and anemic conditions.
Delivery information: See datasheet for contents information.
Documents
TDS Tallquist Hemoglobin Scale
Datasheet
Specifications
- Description:Tallquist Hemoglobin Scale