ContactUsLinkComponent
Swift 2250 Series Budget Microscopes
Swift 2250 Series Budget Microscopes
Catalog # 470039-612
Supplier:  Swift Instruments
undefined
Swift 2250 Series Budget Microscopes
Catalog # 470039-612
Supplier:  Swift Instruments
Supplier Number:  M2251C
Specifications
  • Description:
    Compound Monocular Microscope
  • Environmentally Preferable:
  • Cat. no.:
    470039-612
  • Illumination:
    Cordless LED
  • Supplier No.:
    M2251C
Specifications
About this item
The Swift Budget Compound Microscope is ideal for teaching elementary, middle, and high school microscopy.
  • Full-Size: 34 cm
  • Designed for Intermediate Level Microscopy
  • Rotatable Head
  • Limited Lifetime Manufacturer's Warranty
  • Objectives: 4xD, 10xD and 40xR
It offers 4xD, 10xD and 40xR objectives for 40x–400x total magnification. It boasts many student-proof features including locked-on eyepieces and stage clips, a slip-clutch to avoid gear damage, a cord holder that helps avoid cord pull-out, a convenient mode of cord storage, a bottom plate that protects student hands from electrical shock, and retractable 40X objectives that protect the delicate lenses.
Other safety and value-added aspects include a separate coarse and fine focus, a simple design that is easy to repair, an illuminator diffuser that helps reduce heat to your slides, and a special gear lubricant that remains stable even during normal temperature fluctuations and more.
Ordering information: This product includes a manufacturer's warranty.