Order Entry
Canada
Orders LinkContactUsLinkComponent
Sucrose

Sucrose

Supplier:   Ward's Science
Sucrose
Supplier:  Ward's Science
Certificates
Sucrose

5 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 57-50-1
Formula: C12H22O11
Density: 1.58g/mL
Freezing Point: 185.5°C
Solubility: Water
Synonyms: Cane Sugar, Beet Sugar
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Product Family Options

Product Information

  • Grade
    Size
    Chemical/physical specifications
    Availability
    Price
  • Sucrose
    Laboratory
    500 g
    Crystal
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $29.65
    $29.65
    Each (500g)
       $29.65

    Supplier Number: SS1020-569G
  • Sucrose
    Laboratory
    1 kg
    Crystal
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $34.35
    $34.35
    Each (1kg)
       $34.35

    Supplier Number: SS1020-1KG
  • Sucrose
    Laboratory
    2.5 kg
    Crystal
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $90.20
    $90.20
    Each (2,500g)
       $90.20

    Supplier Number: SS1020-2.5KG
  • 470302-806
    Sucrose, Solution
    Sucrose, Solution
    Laboratory
    120 mL
    20%
    Log in for availability
    Log in for availability
    Each (120ml)
    Each (120ml) - $10.25
    $10.25
    Each (120ml)
       $10.25

    Supplier Number: SS1036-120ML
  • Sucrose
    Reagent
    500 g
    Crystal
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $36.35
    $36.35
    Each (500g)
       $36.35

    Supplier Number: SS1020-500G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.