Student Lab Tables, 1" Phenolic Top
Supplier: Diversified Woodcrafts
Certificates
About this item
Aprons are constructed of solid red oak with heavy-duty 13-gauge steel corner braces. 2.25" square solid oak legs, with ⁵/₁₆" hanger bolts installed, are attached to apron with nuts and washers supplied. They are smoothly machined with a chemical resistant, earth-friendly UV finish.
- Available in 2 heights: sitting height 30", standing height 36"
- 1" phenolic table surface
- Equipped with non-skid adjustable glides for precise leveling and for prevention of rocking on uneven floors
- Black rubber leg boots which offer protection against scuffing and chemical spills
Load capacity: 500 lbs.
Minimal assembly required, legs need to be atttached only.
Certifications: SEFA compliant and MAS Certified Green.
Packaging: Shipped KD with tops attached.
Specifications
- Catalog No:
- 470117-828
- 470117-830
- 470117-836
- 470117-832
- 470117-842
- 470117-838
- 470117-834
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
Specifications
Customers Who Bought This Also Bought