ContactUsLinkComponent
Student Safe-T Compass
Student Safe-T Compass
  470089-478
undefined
Student Safe-T Compass
  470089-478
 :  45759
  • Description:
    Student Safe-T compass
  • Cat. no.:
    470089-478
  • Supplier No.:
    45759
Compass with straight edges made up of shatter-resistant plastic draws circle from 1/4" to 10" in 1/8" diameter increments.
Can be used with pencil or ball-point pen and the straight edge is calibrated inch/metric rules.
 : Supplied with instructions.