Student Platform Scale
Catalog # 470106-426
Supplier: United Scientific Supplies
Specifications
- Cat. no.:470106-426
- Weighing Capacity:11 lbs. (5000 g)
- Supplier No.:KCZ-002-B
Specifications
About this item
Easy for All Grade Levels to Use.
- Constructed of durable, lightweight plastic
- Large read-out front dial
- Clear protective lens fits over the dial
This desktop scale allows you to measure items up to 5000 grams. Display is in pounds and kilograms.