Student Hardware Assortment
Product Details & Documents
Ideal Selection for One Lab Station in a General Chemistry Laboratory.
- For 1 lab station (4 users per set)
Delivery information: Student Hardware Assortment includes the following items: 2 Alligator Clips with 61 cm wire leads; 1 Buret Brush; 1 Buret Clamp, double round jaws; 1 Clamp, utility; 2 Electrodes, carbon rods; 1 Metric Ruler, 15 cm, plastic; 1 Pencil, glass marking, black; 1 Pinch Clamp, Mohr's, 7 cm; 1 Platinum Wire in glass handle; 1 Ring Stand, 51 cm; 1 Ring Support, with clamp; 1 Rubber Tubing, 6.4 mm ID, 1.5 m; 2 Stopper Sets, rubber: #0, 1 hole; #3, 1 hole; #6, 1 hole; #6, 2 hole; 1 Test Tube Brush, 76×13 mm; 2 Test Tube Holders; 1 Test Tube Rack, 12 holes, 5 pins; 1 Thermometer, -20...150 °C; 1 Tongs, crucible, 23 cm; 1 Triangle, 3.8 cm; 1 Wash Bottle, polyethylene, 250 ml; 1 Water Reservoir, pneumatic trough; 1 Wire Gauze, 10×10 cm with fiber center.
Specifications
- Description:Student Hardware Assortment, 1 Lab Station