Order Entry
Canada
Orders LinkContactUsLinkComponent
Student Breadboard

Student Breadboard

Student Breadboard
Catalog # 470110-262
Supplier:   Wards
Supplier #:  CP32742-00
Student Breadboard
Catalog # 470110-262
Supplier:  Wards
Student Breadboard
Student Breadboard

Student Breadboard

Price
$36.30
Log in for availability
Each
Product Details & Documents

This versatile and expandable Student Breadboard is perfect for introductory circuit courses.

  • Board Features 168 Terminal Clips
  • Clips Have 840 Tie Points
  • Perfect for Introductory Circuit Courses
  • Length: 16.5 cm
  • Width: 5.6 cm

The board layout combines bus strips with the breadboard and contains 168 terminal clips offering 840 tie points. Each clip contacts directly inserted wires with diameters between 0.25 mm and 0.84 mm, and the board accepts up to eight 16-pin ICs. A thick Mylar backing eliminates push-through short circuits.

Four through holes allow the board to be attached to a base. The breadboard is 16.5 x 5.6 cm.

Specifications
  • Description:
    Student Breadboard
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.