Structural Adaptations to Water Availability Slide Set
Structural Adaptations to Water Availability Slide Set, Monocots, 10
Catalog # 470183-448
Supplier: Avantor
Jump to:
Product Details & Documents
Examples of Plant Structures from Typical Mesophytes, Xerophytes, and Hydrophytes
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are cross-section unless otherwise specified.
Ordering information: Set includes Monocots: Mesophyte: Zea, mature root; Zea, stem; Zea, leaf; Zea, kernel and embryo (ls); Xerophyte: Yucca, root; Yucca, stem; Yucca, leaf; Yucca, seed (sect); Hydrophyte: Elodea, root or stem; and Elodea, leaf (sect).
Specifications
- Description:Structural Adaptations to Water Availability Slide Set, Monocots
- No. of slides:10