Streptococcus sanguinis
Normal flora of the mouth and throat. Lancefield group H, alpha hemolysis. Optochin resistant. May cause infections in humans. Nonmotile. Cocci; chains. Gram positive reaction. Aerobic. Incubation temperature 37°C. Blood agar growth medium. Note: Pathogen.
Packaged in a 16 x 125 mm tubes. Call our Customer Service for availability of demo plates and other special orders.
- Biosafety Level 2
- Canada: Containment Level 2 - PHAC License required
- Description:Streptococcus sanguinis
- Packaging:Tube
- Media:Blood agar
- Incubation conditions:37 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



