Orders LinkContactUsLinkComponent
Stream Table Recirculating Pump
Stream Table Recirculating Pump
  470007-780
 :   Avantor
Stream Table Recirculating Pump
  470007-780
 :  Avantor
Stream Table Recirculating Pump
Stream Table Recirculating Pump
Stream Table Recirculating Pump
$316.00
Each Pallet of 36
Regulated, Constant Supply of Water
Much easier and less messy than adding water by hand, the 1/70 hp pump circulates up to 300 gallons per hour. It comes with a reservoir and mounting bracket. 115 VAC, 60 Hz. Reservoir size: 14"L x 12"W x 4"H.
  • Description:
    Stream Table Recirculating Pump