$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Foodservice Supplies
Straws, Plastic, Pkg/100, Wrapped
Straws, Plastic, Pkg/100, Wrapped
470012-740
:
INDUSTRIAL SUPPORT INC.
Small Business Enterprise
Straws, Plastic, Pkg/100, Wrapped
470012-740
:
INDUSTRIAL SUPPORT INC.
:
1502 CLEAR
Small Business Enterprise
Description:
Straws, Wrapped
Length:
19.7 cm
Material:
Plastic
Size:
100/pack
Environmentally Preferable:
Cat. no.:
470012-740
Supplier No.:
1502 CLEAR
Diameter:
0.58 cm