Order Entry
Canada
ContactUsLinkComponent
Straws, Plastic
Straws, Plastic
Catalog # 470057-820
Small Business Enterprise
Straws, Plastic
Catalog # 470057-820
Supplier Number:  1510 CLEAR
Small Business Enterprise

Specifications

  • Description:
    Straws
  • Length:
    19.7 cm
  • Material:
    Plastic
  • Size:
    15/pack
  • Environmentally Preferable:
  • Cat. no.:
    470057-820
  • Supplier No.:
    1510 CLEAR
  • Diameter:
    0.58 cm

Specifications

About this item

Straws made of plastic.