Epithelium, Composite Slide
Catalog # 470177-560
Supplier: Avantor
Specifications
- Description:Epithelium, Composite Slide
- Cat. no.:470177-560
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470177-560
- Orientation:Section
Specifications
About this item
Compare and contrast epithelium types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Sections of stratified squamous, columnar, and cuboidal epithelium on the same slide.