Stomates/Guard Cells Slide
Catalog # 470182-512
Supplier: Avantor
Specifications
- Description:Stomates/Guard Cells Slide
- Cat. no.:470182-512
- Stain:Acid-fuchsin and Fast Green
- Supplier No.:WARD470182-512
- Orientation:Whole mount
Specifications
About this item
Pore within the epidermis of plants
- Identifying features clearly distinguishable
- Rigorous quality control standards
Leaf epidermis.