Stomach, Composite Slide
Catalog # 470182-856
Supplier: Avantor
Specifications
- Description:Stomach, Composite Slide
- Cat. no.:470182-856
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-856
- Orientation:Section
Specifications
About this item
Compare and contrast sections of this digestive organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Three regions: cardiac, fundic, and pyloric.