ContactUsLinkComponent
Sterile Scalpel
  470145-792
undefined
Sterile Scalpel
  470145-792
 :  9422
  • Description:
    Sterile Scalpel, 6 1/2" No. 22 Blade
  • Length:
    165 mm (6.5")
  • Material:
    Plastic
  • Environmentally Preferable:
  • Cat. no.:
    470145-792
  • Blade No.:
    #22
  • Supplier No.:
    9422
The permanently fitted blades are durable enough to be reused in non-sterile applications, sterilized before the next use, or simply disposed of for convenience.
They come packaged in sealed wrap to protect them from contamination.