Order Entry
Canada
ContactUsLinkComponent
 
Steam Generator
Catalog # 470149-000
undefined
Steam Generator
Catalog # 470149-000
Supplier Number:  P20220A

Specifications

  • Description:
    Steam Generator
  • Cat. no.:
    470149-000
  • Supplier No.:
    P20220A

Specifications

About this item

Best seller!

  • Volume of water approx. 1.5L
  • Total height with stand approx. 19"
  • Steam Generator height: 8.5", diam. 4.5" approx.

Excellent for experiments in heat and as a source of steam, the seamless, all-aluminum construction of this generator means it won’t collapse even if boiled dry. The durable walls are 1.5 mm thick, and the risk of corrosion is very slim.