Orders LinkContactUsLinkComponent
Staphylococcus epidermidis Slide
Staphylococcus epidermidis Slide, Smear, Gram (+)
  470177-412
 :   Avantor
Staphylococcus epidermidis Slide
  470177-412
 :  Avantor
Staphylococcus epidermidis Slide
Staphylococcus epidermidis Slide
Staphylococcus epidermidis Slide, Smear, Gram (+)
$10.25
Each
Common bacteria on skin
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Spheres occurring singly. Pathogenic on human skin.
  • Description:
    Staphylococcus epidermidis Slide
  • Stain:
    Gram (+)
  • Orientation:
    Smear
Accessories
  •  
    Description
    Media
    Incubation Conditions
  • 470176-542
    Staphylococcus epidermidis
    Tryptic soy agar
    37 °C
    Each - $22.05