Square Polystyrene Disposable, Sterile Petri Dishes
Catalog #: CA25378-115
Supplier: Simport Scientific
Specifications
- Description:Square Polystyrene Disposable, Sterile Petri Dishes
- Form:Liquid
- Cat. no.:CA25378-115
- Supplier No.:D210-16
Specifications
About this item
Petri dishes having a raised square grid on the bottom part. The grid consists of 36 small squares; each square is identified to facilitate special type counting or location of specimens. Supplied with top and bottom in sterile shelf packages. Size: 100 x 15mm.
Customers Who Bought This Also Bought