ContactUsLinkComponent
pinosaurus sp. (Cretaceous) Fossil Specimen
pinosaurus sp. (Cretaceous) Fossil Specimen
Catalog # 470154-802
Supplier:  ADAM MOORE MO
pinosaurus sp. (Cretaceous) Fossil Specimen
Catalog # 470154-802
Supplier:  ADAM MOORE MO
Supplier Number:  TUCSON0056
Specifications
  • Description:
    Spinosaurus sp. (Cretaceous) Fossil Specimen
  • Cat. no.:
    470154-802
  • Supplier No.:
    535140
Specifications
About this item
Well-preserved individual teeth from what may be the largest known carnivorous dinosaur found to date in the fossil record. Exceptional quality and detail. Average size: 1 1/2" L x 1/2" W. Morocco.