Order Entry
Canada
ContactUsLinkComponent
Somso® Median Section of the Head
Somso® Median Section of the Head
Catalog # 470007-334
Supplier:  MARCUS SOMMER
undefined
Somso® Median Section of the Head
Catalog # 470007-334
Supplier:  MARCUS SOMMER
Supplier Number:  BS 6/1

Specifications

  • Description:
    Median section of head
  • Height:
    32 cm
  • Length:
    4.5 cm
  • Cat. no.:
    470007-334
  • Width:
    25.5 cm
  • Supplier No.:
    BS 6/1

Specifications

About this item

Somso® premium quality models.

  • Five year warranty

Natural size, in SOMSO-Plast®. Not detachable. Mounted on a green board.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.