Somso® Half-Size Torso Without Head
Product Details & Documents
Explore The Musculature And Structure Of Major Internal Organs
Students will gain great anatomical insight as they explore the musculature and structure of major internal organs with nine removable parts. The thoracic and abdominal walls are cut away, giving students a view of the underlying viscera, while accurate colors and other functional details help students correlate textbook studies to real life. Designed for introductory lessons to human biology, this model includes a removable heart (two parts), liver with gall bladder, stomach (two parts), lungs, duodenum, kidneys and small and large intestine.
Documents
Somso Production Film
Videos
Specifications
- Description:Half Size Torso Without Head
- No. of keyed items:53
- No. of parts:12