Order Entry
Canada
ContactUsLinkComponent
Somso® DNA Double Helix Model
Somso® DNA Double Helix Model
Catalog # 470029-866
Supplier:  MARCUS SOMMER
undefined
Somso® DNA Double Helix Model
Catalog # 470029-866
Supplier:  MARCUS SOMMER
Supplier Number:  ZOS 57/20

Specifications

  • Description:
    DNA Double Helix
  • Height:
    47.5 cm
  • Length:
    18 cm
  • Cat. no.:
    470029-866
  • Width:
    18 cm
  • Supplier No.:
    ZOS 57/20
  • No. of Parts:
    1

Specifications

About this item

Based On Data From X-Ray Diffraction Studies

  • Mounted On A Rotating Stand For Easy Viewing
  • 5 Year Manufacturers Warranty

Highly accurate in its detail, this model shows a section of a DNA double helix with 10.5 base pairs per turn. The intricate molecular structure of DNA’s major components and the hydrogen bonds between complementary bases are represented by the colored stick presentation in the model’s middle section. Adding further precision, the lower and upper sections of the model are based on the van der Waals radii of the atoms composing the surface.