ContactUsLinkComponent
LaMotte® Soil Sampling Bags
LaMotte® Soil Sampling Bags
  470123-382
 :  LaMotte
undefined
LaMotte® Soil Sampling Bags
  470123-382
 :  LaMotte
 :  0615-J
  • Description:
    Soil sampling bags
  • Size:
    15×10 cm (6×4")
  • Environmentally Preferable:
  • Cat. no.:
    470123-382
  • Supplier No.:
    0615-J
Sealable soil sampling bags have instructions for collection and preparation of soil samples printed directly on each bag.
  • Pack of 100 bags
By preventing contamination or accidental mixing of different samples, these convenient soil bags help insure accurate test results.