Snellen's Eye Test Charts
Catalog # 470111-318
Supplier: Able Printing Service
Specifications
- Width:17.75 cm
- Description:Snellen's Charts
- Length:36 cm
- Cat. no.:470111-318
- Finish:Unlaminated
- Supplier No.:3539
Specifications
About this item
This sturdy chart tests for astigmatism, myopia, or hyperopia.
- Printed in black on heavy white card stock
The chart has an astigmatism test pattern on top and a series of letters in graduated sizes with distance scales for determining myopia or hyperopia. Packaged in a set of six.