Order Entry
Canada
ContactUsLinkComponent
Smedley-Type Hand Dynamometer, FABRICATION ENTERPRISES INC.
Smedley-Type Hand Dynamometer, FABRICATION ENTERPRISES INC.
  470019-104
undefined
Smedley-Type Hand Dynamometer, FABRICATION ENTERPRISES INC.
  470019-104
 :  12-0281

 

  • Description:
    Smedley-Type Hand Dynamometer
  • Height:
    10 cm
  • Length:
    33 cm
  • Environmentally Preferable:
  • Cat. no.:
    470019-104
  • Width:
    20 cm
  • Supplier No.:
    12-0281

 

 

Features Adjustable Hand Stirrup

The dual scale in both English and metric units measures grip strength from 0–220 pounds in 5 pound increments and 0–100 kilograms in 2 kilogram increments. The instrument comes in a protective carrying case.