Skin and Appendages Slide Set
Catalog # 470183-606
Supplier: Avantor
Specifications
- Type:Animal
- Description:Skin and Appendages Slide Set
- No. of Slides:9
- Cat. no.:470183-606
- Supplier No.:WARD470183-606
Specifications
About this item
Illustrates the Cellular Structure of Mammalian Skin and Tissues
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Skin of Hairy Mammal (sect); Mammary Gland, active (sect); Mammary Gland, inactive (sect); Adult Skin, white (sect); Skin, pigmented (sect); Adult Scalp, white (vs); Axillary Skin (sect); Skin (sect); and Eyelid (sect).