Orders LinkContactUsLinkComponent
Single Eye Loupe
Single Eye Loupe
Single Lens Loupe
Catalog # 470021-808
Supplier:   American Scientific
Supplier #:  4402-00
Single Eye Loupe
Catalog # 470021-808
Single Eye Loupe
Single Eye Loupe
Single Lens Loupe
Price
$22.70
Log in for availability
Each
Product Details & Documents
Ideal for Hands-Free Examination of Small Objects
The 5X lens in a plastic frame fits comfortably over one eye, providing magnification when you need it without interrupting your experiment. Focal length: 2 1/2". Size: 1"L x 1 3/8" widest diameter.
Specifications
  • Magnification:
  • Description:
    Single Lens Loupe