To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Single edge razor blade, bulk pack
- Size:0.009" (0.2286 mm)
- Cat. no.:470346-870
- Packaging:Re-closeable clamshell
Specifications
About this item
This 0.009" high carbon surgical steel blade has a non-rust aluminum back.
- Each blade is made of high-carbon surgical steel and non-rust aluminum backing
- Smaller pack of 10 blades comes with safety dispenser for safe usage
- Bulk pack comes in a re-closeable clamshell pack for safe and convenient storage and each blade is individually wrapped and protected in shelling paper
Made with precision honed steel edges with aluminum backing, these blades work for everything from removing paint to peeling stickers from windows.