Order Entry
Canada
ContactUsLinkComponent
Simple Melting Point Apparatus
Simple Melting Point Apparatus
Catalog # 470001-906
Supplier:  Wards
undefined
Simple Melting Point Apparatus
Catalog # 470001-906
Supplier:  Wards
Supplier Number:  CP31777-00

Specifications

  • Description:
    Simple Melting Point Apparatus
  • Cat. no.:
    470001-906
  • Supplier No.:
    31777-00

Specifications

About this item

Easily Measure Melting Point Up To 250° C

Now you can safely demonstrate and measure changes in states of matter in your classroom and laboratory. This economical apparatus consists of an aluminum alloy cylinder, which features a slanted hole drilled in the top to hold thermometers. Simply place your sample on the top of the cylinder as it heats over a bunsen burner. The thermometer allows you to record from a safe distance, the temperature at which the substance melts. Size: 2 1/2" dia. x 2" H.