Orders LinkContactUsLinkComponent
Simple Machines Set
Simple Machines Set
Simple Machines Set
Catalog # 470017-812
Supplier:   Learning Resources
Supplier #:  LER2442
Simple Machines Set
Catalog # 470017-812
Supplier:  Learning Resources
Simple Machines Set
Simple Machines Set
Simple Machines Set
Price
$117.30
Log in for availability
Each
Product Details & Documents
Build 5 simple machines simultaneously!
  • Simple machines
  • Explore concepts of movement
  • Easy assembly
Students can build a lever, pulley, inclined plane, wheel and axle, and wedge all at the same time with this set. These simple machines will enable students to explore effort, force, load, motion and distance as students modify machines with four included weights (two 5g and two 10 g) and eight included rubber bands. Students will develop a deeper understanding of how these five simple machines make work easier. There are labeled pieces which assemble easily and can be separated for storage. Set of 63 pieces includes Activity Guide. Grades 4+.
Caution: WARNING: CHOKING HAZARD - Small parts. Not for children under 3 years.
Specifications
  • Description:
    Simple Machines Set