ContactUsLinkComponent
Fundamentals of Physics Simple Machines
Fundamentals of Physics Simple Machines
Catalog # 470014-336
Supplier:  Wards
undefined
Fundamentals of Physics Simple Machines
Catalog # 470014-336
Supplier:  Wards
Supplier Number:  GLVER
Specifications
  • Description:
    FUNdamentals of Physics: Simple Machines
  • Cat. no.:
    470014-336
  • Supplier No.:
    GLVER
Specifications
About this item
Allow students to investigate the advantage behind different gear ratios.
  • The unique stop system of this device allows students to load masses on the lever arm without it falling
Allow students to investigate the advantage behind different gear ratios. Experimenting with different setups, students discover the mechanical advantage of gears and three types of levers. You can also challenge students to devise a way to lift a mass using the least amount of force.
You can also challenge students to devise a way to lift a mass using the least amount of force.
Accessories information:
Activity requires the fun stand, hooked masses, and spring scales, available separately.
Ordering information:
Kit includes four gears, two levers, mounting hardware, and teacher and student guides.