Silver Oxide
Product Details & Documents
CAS Number: 20667-12-3
Formula Weight: 231.74
Formula: Ag2O
Hazard Info: Oxidizer
Density (g/mL): 7.22
Solubility: Ammonium Hydroxide, Nitric Acid, Sodium Thiosulfate Solution, Water
Synonyms: Disilver Oxide, Silver(I) Oxide
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Documents
Specifications
Specifications
- Description:Silver Oxide
- Size:25 g
- Chemical/physical specifications:Powder
- Grade:Laboratory
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



