Order Entry
Canada
Orders LinkContactUsLinkComponent
Silver Chloride

Silver Chloride

Silver Chloride, Reagent, 25 g
Catalog # 470302-372
Supplier:   Ward's Science
Supplier #:  SS0155-25GR
Silver Chloride
Catalog # 470302-372
Supplier:  Ward's Science
CAS Number:  
Silver Chloride
Silver Chloride

Silver Chloride, Reagent, 25 g

CAS Number:  
Price
408,25 $
Log in for availability
Each (25g)
 
Product Details & Documents

CAS Number: 7783-90-6
Formula Weight: 143.32
Formula: AgCl
Hazard Info: Toxic
Density (g/mL): 5.56
Boiling Point (°C): 1550
Freezing Point (°C): 455
Solubility: Water, Ammonium Hydroxide, Sulfuric Acid, Sodium Thiosulfate and Potassium Bromide Solution
Synonyms: Chloride Salt of Silver
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Silver Chloride
  • Size:
    25 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent

Specifications