Silver Acetate
Silver Acetate, Laboratory, 25 g
Supplier: Ward's Science
Supplier #: SS0158-25G
Jump to:
Product Details & Documents
CAS Number: 563-63-3
Formula: CH3COOAg
Density: 3.259 g/mL
Solubility: Hot Water and Nitric Acid
Synonyms: Argentous Acetate
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is toxic.
Documents
Specifications
Specifications
- Description:Silver Acetate
- Size:25 g
- Chemical/physical specifications:Powder
- Grade:Laboratory
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



