Orders LinkContactUsLinkComponent
Shower Tester
Shower Tester
Shower Tester, max shower head diameter 8"
Catalog # 470105-050
Supplier #:  02-5700-01
Shower Tester
Catalog # 470105-050
Shower Tester
Shower Tester
Shower Tester, max shower head diameter 8"
Price
$304.15
Log in for availability
Each
Product Details & Documents
You know the importance of making sure your safety shower is tested regularly, so it's ready to use in the event of an emergency.
Simply attach this lightweight, PVC coated nylon test funnel to the shower head, and the hemmed bottom passes the water directly and neatly into the bucket.
Will accomodate shower heads up to 8" in diameter. (Bucket not included.)
Specifications
  • Description:
    Shower Tester, max shower head diameter 8"