Microscope Slide, Double Shallow Depression
Double shallow depression slides, 1.5 mm, 3×1"
Catalog # 470003-480
Supplier: PRECISION MOULDED POLYMERS LIMITED
Supplier #: 1350611040004
Microscope Slide, Double Shallow Depression
Catalog # 470003-480
Supplier: PRECISION MOULDED POLYMERS LIMITED
Jump to:
Product Details & Documents
Use this slide to examine liquids more easily
- Quality slide making supplies
- Examine liquids and water borne organisms
Excellent for holding liquids and cultures for microscopic examination. The depressions are approximately 13 mm in diameter and are 0.5-0.8 mm deep.
Specifications
- Description:Double shallow depression slides
- L×W:3×1"
- Thickness:1.5 mm



