Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Separatory Funnels, Squibb

PYREX® Separatory Funnels, Squibb

125 ml, 2 mm
Catalog # 470211-548
Supplier:   Corning
Supplier #:  6402-125
PYREX® Separatory Funnels, Squibb
Catalog # 470211-548
Supplier:  Corning
PYREX® Separatory Funnels, Squibb
PYREX® Separatory Funnels, Squibb

125 ml, 2 mm

Price
$254.00
Log in for availability
Pack of 1 Case of 4
Product Details & Documents

Designed for Long Life and Maximum Safety

  • Heavy walled, borosilicate glass for maximum mechanical strength
  • HPDE standard taper stopper no.:22
  • Replaceable PTFE stopcock assembly

These economical PYREX® separatory funnels are designed to reduce replacement costs by providing replaceable components. The replaceable Polytetrafluorethylene (PTFE) Product Standard stopcock assembly has a simple screw thread locking nut and collar which ensure that the stopcock cannot fall out in use. The stem tip is also replaceable. Its ground end provides a better grip and helps prevent the tip from falling out.

Documents

Specifications
  • Capacity:
    125 ml
  • Stopcock size:
    2 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.