ContactUsLinkComponent
BD BBL Sensi Disc™ Antibiotic Disks
BD BBL Sensi Disc™ Antibiotic Disks
  470201-682
 :  BD
 :  
undefined
BD BBL Sensi Disc™ Antibiotic Disks
  470201-682
 :  BD
 :  260457
 :  
  • Description:
    Stainless Steel Dispenser
  • Cat. no.:
    470201-682
  • Supplier No.:
    260457
Easily observe the sensitivity of various bacteria and fungi to an array of antibiotics with these convenient kits.
  • Convenient
  • Reliable
  • Demonstrate principles of antibiotic resistance and susceptibility
Each magazine contains 50 discs of each type of antibiotic; all you need to do is place them on the bacterial culture and observe the results.  Determine susceptibility or resistance of an organism to specific antibiotics.
A disc dispenser, required, is available separately. Dispenser for Sensi-Disc™ releases one antibiotic disc at a time from a Sensi-Disc™ magazine.