Orders LinkContactUsLinkComponent
Self-Contained Eye Wash Station
Self-Contained Eye Wash Station
Supplier:  
Certificates
Self-Contained Eye Wash Station
0 Options Available Below
Product Details & Documents
Designed for Quick Use Where Potable Water is Not Available.
  • Gravity-fed
  • Easy to see, easy to use
  • The high unit can be placed on a shelf or mounted to a wall
The highly visible, bright yellow polyethylene station is easy to locate in an emergency. Pull the pivot tray down, and within one second the water supply reaches the dual stream heads. A specifically formulated water treatment additive is supplied to prevent the growth of harmful organisms.
Certifications: This unit meets ANSI Z358.1-1981 specifications for both a 15 minute portable eye wash and for a five minute personal eye wash.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470202-578
  • 470016-910
  • Depth:
  • 9"
  • Width:
  • 18"
  • Height:
  • 25"
Specifications
Product Family Options