ContactUsLinkComponent
Screw Lock Scalpel
  470005-456
undefined
Screw Lock Scalpel
  470005-456
 :  140705
  • Description:
    Screw Lock Scalpel
  • Length:
    152 mm (6")
  • Material:
    Nickel Plated
  • Cat. no.:
    470005-456
  • Blade No.:
    #22
  • Supplier No.:
    140705
A versatile, inexpensive scalpel with a safety lock to hold the blade securely, yet allow for simple, safe changes.
The handle is fluted for a better grip, with a blade best suited for heavy dissections. The blade is held in place using a pin and screw spaced for blades fitting a number 4 scalpel.