Screw Compressor Clamp with Closed Jaw
Catalog # 470144-828
Supplier: United Scientific Supplies
Specifications
- Description:Screw Compressor Clamp with Closed Jaw, Brass, Max. Jaw Opening, mm: 19
- Cat. no.:470144-828
- Supplier No.:HSCCL1
Specifications
About this item
Used to control flow in flexible tubing.
- Fits tubes up to 19 mm
- Easy to use thumbscrew
This screw compressor clamps and holds flexible tubes securely closed.