ContactUsLinkComponent
Ward's® Chemistry Colors of a pH Indicator Lab Activity
Ward's® Chemistry Colors of a pH Indicator Lab Activity
Catalog # 470018-100
Supplier:  Avantor
undefined
Ward's® Chemistry Colors of a pH Indicator Lab Activity
Catalog # 470018-100
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ward's® Chemistry Colors of a pH Indicator Lab Activity
  • No. of Lab Groups:
    12
  • Cat. no.:
    470018-100
  • Includes:
    Teacher's Guide/Student Copymasters
  • Supplier No.:
    WARD470018-100
Specifications
About this item
Simple Yet Effective Demonstration
  • Students become more familiar with acid/base chemistry
  • Involves calculations involving dilutions, molarity, pH, and pOH
  • Requires less that 40 minutes to complete
  • Recommended for Grades 7-12
This lab activity helps students become familiarized with the concepts of molarity, dilutions, indicators and PH. Start off with 1 M solutions and then dilute them from a pH of 0 to 14 to a pH of 7. The colors of the solutions will change accordingly as the acidity is altered.
Ordering information: Materials include Hydrochloric acid, Sodium hydroxide, universal indicator. Materials required but not included: test tubes, test tube rack, graduated cylinders, DI water, pipets.
Delivery: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Caution: Hazardous materials.